Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00020214.mm.1 |
TC0X00003230.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020214.mm.1 |
NM_020020.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020215.mm.1 |
TC0X00003230.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020215.mm.1 |
NM_020020.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020213.mm.1 |
TC0X00003230.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020213.mm.1 |
NM_020020.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020212.mm.1 |
TC0X00003230.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020212.mm.1 |
NM_020020.4 |
Refseq |