Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400006755.mm.1 |
TC1400000825.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006755.mm.1 |
NM_009119.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006755.mm.1 |
NM_009119.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006762.mm.1 |
TC1400000825.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006762.mm.1 |
NM_001142441.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006762.mm.1 |
NM_009119.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006762.mm.1 |
NM_009119.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006759.mm.1 |
TC1400000825.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006759.mm.1 |
NM_001142441.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006759.mm.1 |
NM_009119.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006759.mm.1 |
NM_009119.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006756.mm.1 |
TC1400000825.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400006756.mm.1 |
NM_001142441.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006756.mm.1 |
NM_009119.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400006756.mm.1 |
NM_009119.3 |
Refseq |