Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207730_mH
          See other TCP10C GE Assays ›
          Gene Symbol:
          Tcp10c Tcp10a Tcp10b
          Gene Name:
          t-complex protein 10c
          t-complex protein 10a
          t-complex protein 10b
          Gene Aliases:
          D17Leh66C, D17Leh66ca, Gm9880, T66C-a, Tcp-10c
          D17Leh66A, D17Leh66aa, Gm10326, T66A-a, Tcp-10a
          D17Leh66B, D17Leh66ba, T66B-a, Tcp-10b, Tcp-10bt
          Chromosome Location:
          Chr.17: 13354572 - 13377223 on Build GRCm38
          Chromosome Location:
          Chr.17: 7324656 - 7345860 on Build GRCm38
          Chromosome Location:
          Chr.17: 13061109 - 13082226 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm04207730_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcp10c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167578.1 NP_001161050.1 10 - 11 1371 80
          GenBank mRNA AK076614.1 - 10 - 11 1317 80
          AK132653.1 - 11 - 12 1467 80
          BC061173.1 - 10 - 11 1342 6772123 80
          Gene Symbol: Tcp10a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011553.4 NP_035683.2 10 - 11 1271 80
          NR_046291.1 10 - 11 1423 80
          GenBank mRNA AK161455.1 - 10 - 11 1242 80
          BC050771.1 - 10 - 11 1237 6773681 80
          BC052353.1 - 10 - 11 1386 6773668 80
          M22600.1 - 9 - 10 1119 80
          X58169.1 - 10 - 11 1137 80
          X58171.1 - 10 - 11 1244 80
          Gene Symbol: Tcp10b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009341.2 NP_033367.2 10 - 11 1277 80
          GenBank mRNA AK077165.1 - 10 - 11 1286 80
          BC141264.1 - 9 - 10 1148 9055876 80
          M22599.1 - 9 - 10 1269 80
          M22601.1 - 9 - 10 1181 80
          X58170.1 - 9 - 10 1903 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000073.mm.1, TC1700002766.mm.1, TC1700002767.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcp10c
          Click here to find siRNAs for Gene Tcp10a
          Click here to find siRNAs for Gene Tcp10b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700001173.mm.1 TC1700002766.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001173.mm.1 NM_009341.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001173.mm.1 NM_009341.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000573.mm.1 TC1700000073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000573.mm.1 NM_011553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000573.mm.1 NM_011553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000573.mm.1 NR_046291.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000573.mm.1 NR_046291.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001195.mm.1 TC1700002767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001195.mm.1 NM_001167578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001195.mm.1 NM_001167578.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001194.mm.1 TC1700002767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001194.mm.1 NM_001167578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001194.mm.1 NM_001167578.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0