Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04207950_g1
          See other ACKR1 GE Assays ›
          Gene Symbol:
          Ackr1
          Gene Name:
          atypical chemokine receptor 1 (Duffy blood group)
          Gene Aliases:
          AA162249, CCBP1, CD234, Darc, Dfy, ESTM35, FY, GPD
          Chromosome Location:
          Chr.1: 173331886 - 173333503 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04207950_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ackr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010045.2 NP_034175.2 1 - 2 115 102

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_010045.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ackr1

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          signal transduction
          G protein-coupled receptor signaling pathway
          regulation of chemokine production
          chemokine-mediated signaling pathway
          G protein-coupled receptor activity
          chemokine binding
          C-C chemokine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 NM_010045.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AF016584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AF109159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AY289754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AY291295.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 NM_010045.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AF016584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AF109159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AY289754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P110052 AY291295.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331366 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179253 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 NM_010045.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AF016584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AF109159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AY289754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AY291295.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 NM_010045.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AF016584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AF109159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AY289754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P110052 AY291295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 NM_010045.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AF016584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AF109159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AY289754.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AY291295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 NM_010045.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AF016584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AF109159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AY289754.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P110052 AY291295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311611 AF016584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311611 AF109159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311611 AY289754.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311611 AY291295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311611 BC005583.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20837389 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179253 - -
          Whole Mouse Genome G4122A A_51_P311611 AF016584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311611 AF109159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311611 AY289754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311611 AY291295.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311611 BC005583.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 NM_010045.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AF016584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AF109159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AY289754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AY291295.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 NM_010045.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AF016584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AF109159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AY289754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110052 AY291295.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109957 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00444591 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00444545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00444568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00444557 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00444592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20837323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00231462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00910531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00910536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00910532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00910543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00910527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00450434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00531208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00231463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20837376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00444593 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360347 - -
          Affymetrix HT MG-430 PM Array Plate 1432273_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360344 - -
          Mouse Genome 430 2.0 Array 1432273_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432273_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13349_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13349_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100029407.mm.1 TC0100003586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029407.mm.1 NM_010045.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029407.mm.1 NM_010045.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029408.mm.1 TC0100003586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029408.mm.1 NM_010045.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029408.mm.1 NM_010045.2 Refseq
          Mouse Expression Set 430 1432273_a_at - -
          Mouse Expression Set 430 1432273_PM_a_at - -
          Murine Genome U74v2 Set 93412_PM_at - -
          Murine Genome U74v2 Set 93412_at - -
          Mouse Genome 430A 2.0 Array 1432273_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432273_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432273_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432273_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 593117 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3940050 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940050 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2656748 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2656748 - -
          mouse-6 v1.1 (Illumina) scl15911.2_36-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940050 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15911.2_36-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940050 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2656748 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2656748 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl15911.2_36-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix