Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213160_s1
          See other RBM8A GE Assays ›
          Gene Symbol:
          Rbm8a
          Gene Name:
          RNA binding motif protein 8a
          Gene Aliases:
          2310057C03Rik, AA673428, Rbm8
          Chromosome Location:
          Chr.3: 96629928 - 96633791 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213160_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm8a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001102407.1 NP_001095877.1 6 2250 106
          NM_025875.2 NP_080151.2 6 2247 106
          GenBank mRNA AK134935.1 - 1 538 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm8a

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          regulation of alternative mRNA splicing, via spliceosome
          RNA binding
          mRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 CJ075560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 AK009953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 AK010284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 AK082925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 BC020086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 CJ075560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 AK009953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 AK010284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 AK082925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355014 BC020086.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370726 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177978 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 CJ075560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 AK009953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 AK010284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 AK082925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 BC020086.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 CJ075560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 AK009953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 AK010284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 AK082925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355014 BC020086.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22350279 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177978 - -
          Whole Mouse Genome G4122A A_52_P355014 CJ075560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 AK009953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 AK010284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 AK082925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 BC020086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 CJ075560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 AK009953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 AK010284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 AK082925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355014 BC020086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P452578 AK009953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P452578 NM_001102407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P452578 NM_025875.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104736 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02619909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02619914 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02619915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02619912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02619911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02983875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22350279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03138113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03138115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02662109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02635246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02541742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02511019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02983874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02593714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22350267 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494458 - -
          Affymetrix HT MG-430 PM Array Plate 1418119_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418120_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10494452 - -
          Affymetrix Mouse Gene 1.1 ST Array 10360475 - -
          Mouse Genome 430 2.0 Array 1418120_PM_at - -
          Mouse Genome 430 2.0 Array 1418120_at - -
          Mouse Genome 430 2.0 Array 1418119_PM_at - -
          Mouse Genome 430 2.0 Array 1418119_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60365_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60365_PM_at - -
          Mouse Expression Set 430 1418119_at - -
          Mouse Expression Set 430 1418119_PM_at - -
          Mouse Expression Set 430 1418120_at - -
          Mouse Expression Set 430 1418120_PM_at - -
          Murine Genome U74v2 Set 97254_at - -
          Murine Genome U74v2 Set 97254_PM_at - -
          Murine Genome U74v2 Set 129774_f_at - -
          Murine Genome U74v2 Set 168960_PM_at - -
          Murine Genome U74v2 Set 168960_at - -
          Murine Genome U74v2 Set 129774_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1418119_at - -
          Mouse Genome 430A 2.0 Array 1418120_at - -
          Mouse Genome 430A 2.0 Array 1418120_PM_at - -
          Mouse Genome 430A 2.0 Array 1418119_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418120_at - -
          Affymetrix HT Mouse Genome 430A Array 1418119_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418120_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418119_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 540378 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 630168 - -
          MouseWG-6 v2 Expression BeadChip 6220538 - -
          Illumina mouse-6 v1.0 expression beadchip 630168 - -
          Illumina mouse-6 v1.0 expression beadchip 6220538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609615 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2777900 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609615 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2777900 - -
          mouse-6 v1.1 (Illumina) scl060365.6_119-S - -
          mouse-6 v1.1 (Illumina) scl0060365.1_251-S - -
          Illumina Sentrix mouse-6.v1 bead array 630168 - -
          Illumina Sentrix mouse-6.v1 bead array 6220538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0060365.1_251-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl060365.6_119-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220538 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252850 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2609615 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252850 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0060365.1_251-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0