Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213354_s1
          See other ZFAND5 GE Assays ›
          Gene Symbol:
          Zfand5
          Gene Name:
          zinc finger, AN1-type domain 5
          Gene Aliases:
          2310057A04Rik, 5830475F03Rik, AA415485, Za20d2, Zfp216
          Chromosome Location:
          Chr.19: 21271279 - 21286840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213354_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfand5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009551.5 NP_033577.1 1 55 93
          GenBank mRNA AK018054.1 - 1 74 92
          AK076397.1 - 1 56 93
          BC119124.1 - 1 56 8733873 93
          BC119126.1 - 1 56 8733875 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000340.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK076397.1, BC119124.1, BC119126.1, NM_009551.5 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfand5

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          vasculature development
          respiratory system process
          fibroblast migration
          platelet-derived growth factor receptor signaling pathway
          skeletal system morphogenesis
          smooth muscle tissue development
          face development
          DNA binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900003370.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003370.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003370.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003378.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003378.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003378.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003380.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003380.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003380.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003388.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003388.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003388.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003377.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003377.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003377.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003375.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003375.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003375.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003383.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003383.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003383.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003386.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003386.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003386.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003369.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003369.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003369.mm.1 NM_009551.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003371.mm.1 TC1900000340.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003371.mm.1 NM_009551.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003371.mm.1 NM_009551.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 452162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline