Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213972_s1
          See other C1S2 GE Assays ›
          Gene Symbol:
          C1s2
          Gene Name:
          complement component 1, s subcomponent 2
          Gene Aliases:
          C1sb, EG317677, Gm5077
          Chromosome Location:
          Chr.6: 124624625 - 124636085 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213972_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C1s2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173864.2 NP_776289.2 11 1553 83
          GenBank mRNA AF459020.1 - 11 1553 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003109.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C1s2

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          proteolysis
          complement activation
          complement activation, classical pathway
          innate immune response
          serine-type endopeptidase activity
          calcium ion binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P39505 NM_173864.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P39505 AF459020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P39505 NM_173864.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P39505 AF459020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150179 NM_173864.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150179 AF459020.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4285 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150179 NM_173864.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150179 AF459020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P39505 NM_173864.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P39505 AF459020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P39505 NM_173864.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P39505 AF459020.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816194 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06055912 - -
          Whole Mouse Genome G4122A A_52_P39505 NM_173864.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P39505 AF459020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P39505 NM_173864.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P39505 AF459020.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01437485 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05221562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05221560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05221560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05225930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05028690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05221562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05584971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05539439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05755726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05584972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05028691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816141 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547754 - -
          Affymetrix Mouse Gene 1.1 ST Array 10547752 - -
          Affymetrix Mouse Clariom S Array PSR0600024036.mm.1 TC0600003109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024036.mm.1 NM_173864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024036.mm.1 NM_173864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024035.mm.1 TC0600003109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024035.mm.1 NM_173864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024035.mm.1 NM_173864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024032.mm.1 TC0600003109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024032.mm.1 NM_173864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024032.mm.1 NM_173864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024033.mm.1 TC0600003109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024033.mm.1 NM_173864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024033.mm.1 NM_173864.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024034.mm.1 TC0600003109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024034.mm.1 NM_173864.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024034.mm.1 NM_173864.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 537478 - -
          ABI Mouse Genome Survey Microarray 629498 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6660167 - -
          MouseWG-6 v2 Expression BeadChip 1240072 - -
          Illumina mouse-6 v1.0 expression beadchip 1980114 - -
          Illumina mouse-6 v1.0 expression beadchip 6660167 - -
          Illumina mouse-6 v1.0 expression beadchip 1240072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660167 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639513 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260226 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2752720 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639513 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260226 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2752720 - -
          mouse-6 v1.1 (Illumina) scl050908.2_30-S - -
          mouse-6 v1.1 (Illumina) scl00317677.1_326-S - -
          mouse-6 v1.1 (Illumina) scl00317677.1_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980114 - -
          Illumina Sentrix mouse-6.v1 bead array 1240072 - -
          Illumina Sentrix mouse-6.v1 bead array 6660167 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00317677.1_41-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00317677.1_326-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl050908.2_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660167 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1260226 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2929594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2752720 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2639513 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2752720 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2929594 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260226 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639513 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0