Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1440761_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10410973 |
- |
- |
Mouse Genome 430 2.0 Array |
1440761_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1440761_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
TC1300002412.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
NR_015501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
NR_015501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
NR_028313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
NR_028313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
NR_028314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016009.mm.1 |
NR_028314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
TC1300002412.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
NR_015501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
NR_015501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
NR_028313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
NR_028313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
NR_028314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016010.mm.1 |
NR_028314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016004.mm.1 |
TC1300002412.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016004.mm.1 |
NR_015501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016004.mm.1 |
NR_015501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
TC1300002412.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
NR_015501.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
NR_015501.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
NR_028313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
NR_028313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
NR_028314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016011.mm.1 |
NR_028314.1 |
Refseq |
Mouse Expression Set 430 |
1440761_PM_at |
- |
- |
Mouse Expression Set 430 |
1440761_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1440761_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1440761_at |
- |
- |