Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04235657_m1
          See other 4933439C10RIK GE Assays ›
          Gene Symbol:
          4933439C10Rik
          Gene Name:
          RIKEN cDNA 4933439C10 gene
          Gene Aliases:
          2610028M21Rik
          Chromosome Location:
          Chr.11: 59505685 - 59511067 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04235657_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933439C10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015585.2 3 - 4 1430 68
          GenBank mRNA AK011602.1 - 3 - 4 963 68
          AK017115.1 - 3 - 4 1431 68
          AK030920.1 - 1458 68
          AK163429.1 - 3 - 4 1426 68
          AK169756.1 - 3 - 4 1460 68
          BC065402.1 - 2 - 3 136 68
          BC099591.1 - 3 - 4 677 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ928558.1 - 3 - 4 436 Chr. 11 - 59506679 - 59510106 [+] on Build GRCm38 68
          GenBankmRNA BX525008.1 - 3 - 4 343 Chr. 11 - 59506791 - 59510846 [+] on Build GRCm38 68
          GenBankmRNA BY762673.1 - 2 - 3 245 Chr. 11 - 59507190 - 59511067 [+] on Build GRCm38 68
          GenBankmRNA AI036408.1 - 2 - 3 122 Chr. 11 - 59507641 - 59511012 [+] on Build GRCm38 68
          GenBankmRNA BP760795.1 - 2 - 3 Chr. 11 - 59507840 - 59511073 [-] on Build GRCm38 68
          GenBankmRNA EH098200.1 - 1 - 2 104 Chr. 11 - 59507842 - 59511069 [+] on Build GRCm38 68
          GenBankmRNA W98666.1 - 326 Chr. - - [-] on Build 68
          GenBankmRNA BY754733.1 - 2 - 3 309 Chr. 11 - 59507225 - 59510103 [+] on Build GRCm38 68
          GenBankmRNA AI429744.1 - 2 - 3 Chr. 11 - 59507887 - 59511073 [-] on Build GRCm38 68
          GenBankmRNA EH099787.1 - 2 - 3 245 Chr. 11 - 59507189 - 59510930 [+] on Build GRCm38 68
          GenBankmRNA CX243337.1 - 3 - 4 352 Chr. 11 - 59506763 - 59510086 [+] on Build GRCm38 68
          GenBankmRNA BE133382.1 - 2 - 3 136 Chr. 11 - 59507283 - 59510940 [+] on Build GRCm38 68
          GenBankmRNA EH102917.1 - 2 - 3 245 Chr. 11 - 59507189 - 59511035 [+] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger with KRAB and SCAN domains 17 Zkscan17 Nizp1,
          Zfp496,
          Znf496
          -1.2kb Anti-sense Chr. 11 - 59485520 - 59506640 on NCBI Build GRCm38
          Downstream NLR family,
          pyrin domain containing 3
          Nlrp3 AGTAVPRL,
          AII/AVP,
          Cias1,
          FCAS,
          FCU,
          MWS,
          Mmig1,
          NALP3,
          Pypaf1
          31.8kb Sense Chr. 11 - 59541569 - 59566956 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix