Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04236274_m1
          See other GM2762 GE Assays ›
          Gene Symbol:
          Gm2762
          Gene Name:
          predicted gene 2762
          Gene Aliases:
          1700025I17Rik
          Chromosome Location:
          Chr.13: 53393689 - 53455630 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04236274_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm2762
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037991.1 7 - 8 803 74
          GenBank mRNA AK006343.1 - 7 - 8 781 74
          AK030238.1 - 7 - 8 804 74
          BC100495.1 - 7 - 8 784 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700058M13 gene 1700058M13Rik -36.6kb Anti-sense Chr. 13 - 53398086 - 53415880 on NCBI Build GRCm38
          Downstream msh homeobox 2 Msx2 BB122635,
          Hox-8,
          Hox8,
          Hox8.1
          11.3kb Anti-sense Chr. 13 - 53466881 - 53472780 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000618.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359630 BC111581.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359630 NM_146568.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570874 AK035548.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570874 NM_009922.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109475 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07877865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07877880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02693565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02693558 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02693512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02693495 - -
          Whole Mouse Genome G4122A A_52_P2774 AK030238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2774 BC100495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2774 NR_037991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2774 AK006343.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124006 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171807 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02693558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10891087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10998034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29044001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11101345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10951901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10709829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10998039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10709836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10929340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29044003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29043718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10768918 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431157_PM_at - -
          Mouse Genome 430 2.0 Array 1431157_PM_at - -
          Mouse Genome 430 2.0 Array 1431157_at - -
          Affymetrix Mouse Clariom S Array PSR1300003938.mm.1 TC1300000618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300003938.mm.1 NR_037991.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003938.mm.1 NR_037991.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003937.mm.1 TC1300000618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300003937.mm.1 NR_037991.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003937.mm.1 NR_037991.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003939.mm.1 TC1300000618.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300003939.mm.1 NR_037991.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003939.mm.1 NR_037991.1 Refseq
          Mouse Expression Set 430 1431157_PM_at - -
          Mouse Expression Set 430 1431157_at - -
          Murine Genome U74v2 Set 167386_at - -
          Murine Genome U74v2 Set 167386_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431157_at - -
          Affymetrix HT Mouse Genome 430B Array 1431157_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 448228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0