Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04238058_m1
          See other GM10863 GE Assays ›
          Gene Symbol:
          Gm10863
          Gene Name:
          predicted gene 10863
          Gene Aliases:
          ENSMUSG00000075555
          Chromosome Location:
          Chr.15: 79166066 - 79216401 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04238058_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10863
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029470.1 4 - 5 393 69
          GenBank mRNA AK145218.1 - 168 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF302801.1 - 4 - 5 393 Chr. 15 - 79166066 - 79216497 [+] on Build GRCm38 69
          GenBankmRNA BY735273.1 - 2 - 3 168 Chr. 15 - 79209311 - 79217156 [+] on Build GRCm38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SRY (sex determining region Y)-box 10 Sox10 Dom,
          Sox21
          -45.0kb Anti-sense Chr. 15 - 79154909 - 79165235 on NCBI Build GRCm38
          Downstream protein interacting with C kinase 1 Pick1 Prkcabp 12.8kb Sense Chr. 15 - 79229166 - 79249466 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000668.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK145218.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055374 NR_029470.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055374 NR_029470.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079178 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140916 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430925 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430890 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171029 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430047 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430630 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078885 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430393 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08431247 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175371 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078869 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430373 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078433 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078636 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08431170 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079389 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079406 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079215 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08430219 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08429691 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08431199 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078649 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079414 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079456 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03079064 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078990 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171029 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115290 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03078636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03078869 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03079215 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140916 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08430393 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08431170 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30365040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30365043 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500004999.mm.1 TC1500000668.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004999.mm.1 NR_029470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004999.mm.1 NR_029470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004997.mm.1 TC1500000668.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004997.mm.1 NR_029470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004997.mm.1 NR_029470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004998.mm.1 TC1500000668.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004998.mm.1 NR_029470.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004998.mm.1 NR_029470.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0