Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04238519_m1
          See other AA387883 GE Assays ›
          Gene Symbol:
          AA387883
          Gene Name:
          expressed sequence AA387883
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 52923181 - 52926931 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04238519_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AA387883
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030678.1 1 - 2 320 72
          GenBank mRNA AK043996.1 - 1 - 2 321 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ917305.1 - 1 - 2 325 Chr. 19 - 52923194 - 52926129 [+] on Build GRCm38 72
          GenBankmRNA BB641900.1 - 1 - 2 318 Chr. 19 - 52923183 - 52925905 [+] on Build GRCm38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream insulin I Ins1 Ins-1,
          Ins2-rs1
          -657.7kb Sense Chr. 19 - 52264323 - 52265476 on NCBI Build GRCm38
          Downstream X-prolyl aminopeptidase (aminopeptidase P) 1,
          soluble
          Xpnpep1 D230045I08Rik,
          sAMP
          5.3kb Anti-sense Chr. 19 - 52932227 - 53038654 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000744.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103493 AK043996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103493 NR_030678.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14245184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14357323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14215567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14470772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32179447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900006587.mm.1 TC1900000744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900006587.mm.1 NR_030678.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006587.mm.1 NR_030678.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006585.mm.1 TC1900000744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900006585.mm.1 NR_030678.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900006585.mm.1 NR_030678.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0