Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10486201 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200037001.mm.1 |
TC0200004430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037001.mm.1 |
NR_030683.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037001.mm.1 |
NR_030683.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037002.mm.1 |
TC0200004430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037002.mm.1 |
NR_030683.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037002.mm.1 |
NR_030683.1 |
Refseq |