Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239373_m1
          See other BC065397 GE Assays ›
          Gene Symbol:
          BC065397
          Gene Name:
          cDNA sequence BC065397
          Gene Aliases:
          R75336, parl7
          Chromosome Location:
          Chr.X: 136742957 - 136803364 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239373_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC065397
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033324.1 7 - 8 2519 102
          GenBank mRNA AK142310.1 - 7 - 8 2500 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CR758929.1 - 3 - 4 Chr. X - 136758604 - 136802883 [-] on Build GRCm38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mortality factor 4 like 2 Morf4l2 2410017O14Rik,
          Mrgx,
          Sid393p,
          mKIAA0026
          -16.7kb Anti-sense Chr. X - 136732942 - 136743686 on NCBI Build GRCm38
          Downstream proteolipid protein (myelin) 1 Plp1 DM20,
          Plp,
          jimpy,
          jp,
          msd,
          rsh
          60.7kb Sense Chr. X - 136822746 - 136838582 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001336.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 AK142310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 AK160692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 NR_033324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 AK084423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 AK142310.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 AK160692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 NR_033324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1004509 AK084423.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749932 AF057287.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749932 AK029917.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749932 AK048326.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749932 AK049400.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749932 BC031154.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749932 NM_010472.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 AK016945.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 AK029561.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 AK039149.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 AK083793.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 AK140231.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 AY033991.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 BC066156.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 NM_001081456.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444740 NM_148937.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 152575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325775 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555176 AK006500.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168480 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146611 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163959 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167397 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141235 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P793384 AK142310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P793384 BC065397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P793384 NR_033324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P793384 AK142310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P793384 BC065397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P793384 NR_033324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 AK142310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 AK160692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 NR_033324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 AK084423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 AK142310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 AK160692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 NR_033324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1004509 AK084423.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03760200 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146611 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141235 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167397 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03760124 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09430643 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03760185 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163959 - -
          Whole Mouse Genome G4122A A_52_P793384 AK142310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P793384 BC065397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P793384 NR_033324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P793384 AK142310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P793384 BC065397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P793384 NR_033324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 AK142310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 AK160692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 NR_033324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 AK084423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 AK142310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 AK160692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 NR_033324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1004509 AK084423.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110932 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141235 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163959 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14839315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14952762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14608574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14952756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14717817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601883 - -
          Affymetrix HT MG-430 PM Array Plate 1438330_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10601882 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601880 - -
          Mouse Genome 430 2.0 Array 1438330_PM_at - -
          Mouse Genome 430 2.0 Array 1438330_at - -
          Affymetrix Mouse Clariom S Array PSR0X00008431.mm.1 TC0X00001336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008431.mm.1 NR_033324.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008431.mm.1 NR_033324.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008432.mm.1 TC0X00001336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008432.mm.1 NR_033324.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008432.mm.1 NR_033324.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008425.mm.1 TC0X00001336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008425.mm.1 NR_033324.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008425.mm.1 NR_033324.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008430.mm.1 TC0X00001336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008430.mm.1 NR_033324.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008430.mm.1 NR_033324.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008428.mm.1 TC0X00001336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008428.mm.1 NR_033324.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008428.mm.1 NR_033324.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008429.mm.1 TC0X00001336.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008429.mm.1 NR_033324.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008429.mm.1 NR_033324.1 Refseq
          Mouse Expression Set 430 1438330_at - -
          Mouse Expression Set 430 1438330_PM_at - -
          Murine Genome U74v2 Set 166747_at - -
          Murine Genome U74v2 Set 166747_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438330_at - -
          Affymetrix HT Mouse Genome 430B Array 1438330_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 553248 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101230333 - -
          Illumina mouse-6 v1.0 expression beadchip 101230333 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2582348 - -
          Illumina Sentrix mouse-6.v1 bead array 101230333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101230333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2869654 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2869654 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2869658 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0