Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239847_m1
          See other GM16287 GE Assays ›
          Gene Symbol:
          Gm16287
          Gene Name:
          predicted gene 16287
          Gene Aliases:
          ENSMUSG00000073739, OTTMUSG00000009902, OTTMUSG00000033208
          Chromosome Location:
          Chr.4: 139175407 - 139180655 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239847_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16287
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033543.1 2 - 3 1142 106
          GenBank mRNA AK141099.1 - 2 - 3 1143 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitochondrial inner membrane organizing system 1 Minos1 2310028O11Rik -44.3kb Sense Chr. 4 - 139101814 - 139131113 on NCBI Build GRCm38
          Downstream capping protein (actin filament) muscle Z-line,
          beta
          Capzb 1700120C01Rik,
          AI325129,
          CPB1,
          CPB2,
          CPbeat2,
          CPbeta1,
          CPbeta2,
          Cappb1
          13.9kb Anti-sense Chr. 4 - 139192899 - 139291820 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003845.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK141099.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088113 NR_033543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088113 AK141099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088113 NR_033543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088113 AK141099.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03711408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03838723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03837889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03837891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03631718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03203262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03631719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03203258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03203259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03203255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03837893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03848699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03417259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03838726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23350069 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10517687 - -
          Affymetrix Mouse Clariom S Array PSR0400031685.mm.1 TC0400003845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031685.mm.1 NR_033543.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031685.mm.1 NR_033543.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031684.mm.1 TC0400003845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031684.mm.1 NR_033543.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031684.mm.1 NR_033543.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031686.mm.1 TC0400003845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031686.mm.1 NR_033543.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031686.mm.1 NR_033543.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0