Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04239860_m1
          See other GM15348 GE Assays ›
          Gene Symbol:
          Gm15348
          Gene Name:
          predicted gene 15348
          Gene Aliases:
          ENSMUSG00000074469, OTTMUSG00000020606
          Chromosome Location:
          Chr.8: 12706944 - 12719127 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04239860_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15348
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033546.1 3 - 4 570 95
          GenBank mRNA AK143914.1 - 3 - 4 571 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tubulin,
          gamma complex associated protein 3
          Tubgcp3 GCP3,
          Spc98p
          -34.7kb Sense Chr. 8 - 12614277 - 12672243 on NCBI Build GRCm38
          Downstream ATPase,
          class VI,
          type 11A
          Atp11a 4930558F19Rik,
          AU040868,
          Atpc1h,
          Ih
          43.0kb Anti-sense Chr. 8 - 12757016 - 12868728 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001718.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008042 AK143914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008042 NR_033546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008042 AK143914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008042 NR_033546.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07425430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07276772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25663397 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10577093 - -
          Affymetrix Mouse Clariom S Array PSR0800013913.mm.1 TC0800001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013913.mm.1 NR_033546.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013913.mm.1 NR_033546.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013914.mm.1 TC0800001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013914.mm.1 NR_033546.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013914.mm.1 NR_033546.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0