Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04240440_s1
          See other 1700028B04RIK GE Assays ›
          Gene Symbol:
          1700028B04Rik
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 28496941 - 28497492 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04240440_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700028B04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033605.1 1 350 77
          GenBank mRNA AK006445.1 - 549 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leucine rich repeat and fibronectin type III domain containing 1 Lrfn1 BC004018,
          SALM2,
          Semo1
          -28.7kb Sense Chr. 7 - 28451985 - 28468242 on NCBI Build GRCm38
          Downstream interferon lambda 2 Ifnl2 EG330496,
          IL-28A,
          Il28a
          11.3kb Anti-sense Chr. 7 - 28508784 - 28510532 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000503.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P105830 AK006445.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167330 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06149677 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105830 AK006445.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06554226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06208217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06066659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06475121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25022137 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700004094.mm.1 TC0700000503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004094.mm.1 NR_033605.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004094.mm.1 NR_033605.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 337951 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0