Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242134_m1
          See other 5730422E09RIK GE Assays ›
          Gene Symbol:
          5730422E09Rik
          Gene Name:
          RIKEN cDNA 5730422E09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 149119565 - 149182036 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242134_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5730422E09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015478.2 1 - 2 46 73
          GenBank mRNA AK082086.1 - 72 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB596182.2 - 2 - 3 72 Chr. 5 - 149128270 - 149184063 [-] on Build GRCm38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glucosamine-6-phosphate deaminase 1 pseudogene Gm8615 EG667410 Sense Chr. 5 - 149154128 - 149156192 on NCBI Build GRCm38
          Downstream ubiquitin specific peptidase like 1 Uspl1 E430026A01Rik 2.5kb Anti-sense Chr. 5 - 149184560 - 149215434 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003650.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1200000116.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2228710 AK017599.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2228715 AK082086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2228715 NR_015478.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180211 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996290 AK031519.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180211 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01249472 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05787808 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05787793 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01249514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05787825 - -
          Whole Mouse Genome G4122A A_52_P598379 AK017599.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106800 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105833 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180211 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01249514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04808054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04194550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04952378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04800567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04171132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04952368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04194552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04250277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04224445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04577036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24183376 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444601_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454575_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433357_PM_at - -
          Mouse Genome 430 2.0 Array 1454575_at - -
          Mouse Genome 430 2.0 Array 1433357_at - -
          Mouse Genome 430 2.0 Array 1433357_PM_at - -
          Mouse Genome 430 2.0 Array 1454575_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500033545.mm.1 TC0500003650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033545.mm.1 NR_015478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033542.mm.1 TC0500003650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033542.mm.1 NR_015478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033542.mm.1 NR_015478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033543.mm.1 TC0500003650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033543.mm.1 NR_015478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033543.mm.1 NR_015478.2 Refseq
          Mouse Expression Set 430 1433357_PM_at - -
          Mouse Expression Set 430 1444601_PM_at - -
          Mouse Expression Set 430 1454575_at - -
          Mouse Expression Set 430 1433357_at - -
          Mouse Expression Set 430 1444601_at - -
          Mouse Expression Set 430 1454575_PM_at - -
          Murine Genome U74v2 Set 134171_at - -
          Murine Genome U74v2 Set 134171_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433357_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454575_at - -
          Affymetrix HT Mouse Genome 430B Array 1454575_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 825844 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0