Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242495_s1
          See other IL1BOS GE Assays ›
          Gene Symbol:
          Il1bos
          Gene Name:
          interleukin 1 beta, opposite strand
          Gene Aliases:
          9430083I22, Gm14047, OTTMUSG00000015529
          Chromosome Location:
          Chr.2: 129369902 - 129375733 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242495_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1bos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015474.1 2 144 82
          GenBank mRNA AK035070.1 - 2 145 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005484.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P494020 NR_015474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P494020 AK035070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P494020 AK076405.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P494020 NR_015474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P494020 AK035070.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P494020 AK076405.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P494020 NR_015474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P494020 AK035070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P494020 AK076405.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P494020 NR_015474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P494020 AK035070.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P494020 AK076405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494020 NR_015474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494020 AK035070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494020 AK076405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494020 NR_015474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494020 AK035070.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P494020 AK076405.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P01257892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21624534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01257898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21624538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446631_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476012 - -
          Mouse Genome 430 2.0 Array 1446631_at - -
          Mouse Genome 430 2.0 Array 1446631_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329514_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329514_at - -
          Affymetrix Mouse Clariom S Array PSR0200015457.mm.1 TC0200005484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015457.mm.1 NR_015474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015457.mm.1 NR_015474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046676.mm.1 TC0200005484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200046676.mm.1 NR_015474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200046676.mm.1 NR_015474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015454.mm.1 TC0200005484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015454.mm.1 NR_015474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015454.mm.1 NR_015474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015462.mm.1 TC0200005484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015462.mm.1 NR_015474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015462.mm.1 NR_015474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015455.mm.1 TC0200005484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015455.mm.1 NR_015474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015455.mm.1 NR_015474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015456.mm.1 TC0200005484.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200015456.mm.1 NR_015474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200015456.mm.1 NR_015474.1 Refseq
          Mouse Expression Set 430 1446631_PM_at - -
          Mouse Expression Set 430 1446631_at - -
          Affymetrix HT Mouse Genome 430B Array 1446631_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446631_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 354261 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238850 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238850 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238850 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix