Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04242656_s1
          See other 9330151L19RIK GE Assays ›
          Gene Symbol:
          9330151L19Rik
          Gene Name:
          RIKEN cDNA 9330151L19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 69197211 - 69199868 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04242656_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330151L19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033222.1 2 2446 76
          GenBank mRNA AK034052.1 - 2 2434 76
          AK161156.1 - 1 2563 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX639899.1 - 1 Chr. 12 - 69199611 - 69199869 [-] on Build GRCm38 76
          GenBankmRNA BY629770.1 - 1 291 Chr. 12 - 69199478 - 69199867 [+] on Build GRCm38 76
          GenBankmRNA BB078170.2 - 1 580 Chr. 12 - 69199190 - 69199861 [+] on Build GRCm38 76
          GenBankmRNA AA855571.1 - 1 Chr. 12 - 69199403 - 69199813 [-] on Build GRCm38 76
          GenBankmRNA AI465095.1 - 1 Chr. 12 - 69199374 - 69199814 [-] on Build GRCm38 76
          GenBankmRNA BB738004.1 - 1 281 Chr. 12 - 69199487 - 69199868 [+] on Build GRCm38 76
          GenBankmRNA AW488232.1 - 1 Chr. 12 - 69199342 - 69199864 [-] on Build GRCm38 76
          GenBankmRNA BI695756.1 - 1 339 Chr. 12 - 69199430 - 69199869 [+] on Build GRCm38 76
          GenBankmRNA BB009905.2 - 1 550 Chr. 12 - 69199219 - 69199867 [+] on Build GRCm38 76
          GenBankmRNA BB265695.1 - 1 202 Chr. 12 - 69199567 - 69199860 [+] on Build GRCm38 76
          GenBankmRNA BF719271.1 - 1 Chr. 12 - 69199457 - 69199835 [-] on Build GRCm38 76
          GenBankmRNA BB739444.1 - 1 296 Chr. 12 - 69199471 - 69199867 [+] on Build GRCm38 76
          GenBankmRNA BY638311.1 - 317 Chr. - - [-] on Build 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mannoside acetylglucosaminyltransferase 2 Mgat2 AA407964,
          CDGS2,
          GLCNACTII,
          GNT-II,
          GNT2
          -11.0kb Sense Chr. 12 - 69184158 - 69186773 on NCBI Build GRCm38
          Downstream polymerase (DNA directed),
          epsilon 2 (p59 subunit)
          Pole2 1.9kb Anti-sense Chr. 12 - 69201773 - 69228242 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000564.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK161156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 AK034052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P120066 NR_033222.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128488 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130902 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130902 - -
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK161156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 AK034052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P120066 NR_033222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P579993 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125567 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130902 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1434166_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10400642 - -
          Affymetrix Mouse Gene 1.1 ST Array 10396076 - -
          Mouse Genome 430 2.0 Array 1434166_PM_at - -
          Mouse Genome 430 2.0 Array 1434166_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 414085_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 414085_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200003754.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003754.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003754.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003762.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003762.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003762.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003765.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003765.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003765.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003752.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003752.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003752.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003758.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003758.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003758.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003763.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003763.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003763.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003764.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003764.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003764.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003768.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003768.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003768.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003755.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003755.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003755.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003760.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003760.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003760.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003757.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003757.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003757.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003761.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003761.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003761.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003767.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003767.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003767.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003756.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003756.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003756.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003759.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003759.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003759.mm.1 NR_033222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003766.mm.1 TC1200000564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003766.mm.1 NR_033222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003766.mm.1 NR_033222.1 Refseq
          Mouse Expression Set 430 1434166_at - -
          Mouse Expression Set 430 1434166_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434166_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434166_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100670670 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256968 - -
          Illumina Sentrix mouse-6.v1 bead array 100670670 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100670670 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256968 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0