Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04243917_mH
          See other AI607873 GE Assays ›
          Gene Symbol:
          AI607873
          Gene Name:
          expressed sequence AI607873
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 173723427 - 173741837 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04243917_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI607873
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204910.1 NP_001191839.1 5 - 6 2548 98
          GenBank mRNA BC150711.1 - 2563 9087622 98
          BE634329.1 - 1 - 2 230 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003593.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          double-stranded DNA binding
          RNA binding
          transcription factor binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P325281 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152660 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116415 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081105 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P325281 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359454 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116415 - -
          Whole Mouse Genome G4122A A_52_P86965 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01049037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00610421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00610435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360378 - -
          Affymetrix HT MG-430 PM Array Plate 1457035_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360377 - -
          Mouse Genome 430 2.0 Array 1457035_at - -
          Mouse Genome 430 2.0 Array 1457035_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226691_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226691_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100029489.mm.1 TC0100003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029489.mm.1 NM_001204910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029489.mm.1 NM_001204910.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029488.mm.1 TC0100003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029488.mm.1 NM_001204910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029488.mm.1 NM_001204910.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029490.mm.1 TC0100003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029490.mm.1 NM_001204910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029490.mm.1 NM_001204910.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029487.mm.1 TC0100003593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029487.mm.1 NM_001204910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029487.mm.1 NM_001204910.1 Refseq
          Murine Genome U74v2 Set 92251_PM_f_at - -
          Murine Genome U74v2 Set 92251_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1457035_at - -
          Affymetrix HT Mouse Genome 430B Array 1457035_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254577 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0