Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04243980_s1
          See other 5430421N21RIK GE Assays ›
          Gene Symbol:
          5430421N21Rik
          Gene Name:
          RIKEN cDNA 5430421N21 gene
          Gene Aliases:
          Krt83
          Chromosome Location:
          Chr.15: 101485130 - 101491512 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04243980_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430421N21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001201323.1 NP_001188252.1 9 1651 97
          GenBank mRNA AI561926.1 - 1 97
          AK133718.1 - 9 1652 97
          M92088.1 - 9 1636 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002259.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          aging
          hair cycle
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P12468510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11853303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12134883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11853305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12363050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11889029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11889033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12134884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12134894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12134907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432707 - -
          Affymetrix HT MG-430 PM Array Plate 1427118_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10432695 - -
          Mouse Genome 430 2.0 Array 1427118_at - -
          Mouse Genome 430 2.0 Array 1427118_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500018569.mm.1 TC1500002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018569.mm.1 NM_001201323.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018569.mm.1 NM_001201323.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018583.mm.1 TC1500002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018583.mm.1 NM_001201323.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018583.mm.1 NM_001201323.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018570.mm.1 TC1500002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018570.mm.1 NM_001201323.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018570.mm.1 NM_001201323.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018576.mm.1 TC1500002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018576.mm.1 NM_001201323.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018576.mm.1 NM_001201323.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018571.mm.1 TC1500002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018571.mm.1 NM_001201323.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018571.mm.1 NM_001201323.1 Refseq
          Mouse Expression Set 430 1427118_PM_at - -
          Mouse Expression Set 430 1427118_at - -
          Murine Genome U74v2 Set 93555_PM_at - -
          Murine Genome U74v2 Set 93555_at - -
          Murine Genome U74v2 Set 169745_PM_f_at - -
          Murine Genome U74v2 Set 169745_f_at - -
          Mouse Genome 430A 2.0 Array 1427118_PM_at - -
          Mouse Genome 430A 2.0 Array 1427118_at - -
          Affymetrix HT Mouse Genome 430A Array 1427118_at - -
          Affymetrix HT Mouse Genome 430A Array 1427118_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 850385 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0