Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04277531_s1
          See other E530001F21RIK GE Assays ›
          Gene Symbol:
          E530001F21Rik
          Gene Name:
          RIKEN cDNA E530001F21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 105164378 - 105165505 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04277531_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E530001F21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002167.2 1 118 103
          GenBank mRNA AK089116.1 - 1 120 105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB609231.2 - 1 117 Chr. X - 105164878 - 105165502 [-] on Build GRCm38 105

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          33605
          Gm33605 Sense Chr. X - 105146942 - 105176031 on NCBI Build GRCm38
          Downstream microRNA 384 Mir384 Mirn384,
          mir-384,
          mmu-mir-384
          178.8kb Sense Chr. X - 105344282 - 105344369 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002809.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162633 AK089116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162633 NR_002167.2 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P14819490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15119426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14598589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32579512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00017802.mm.1 TC0X00002809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017802.mm.1 NR_002167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017805.mm.1 TC0X00002809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017805.mm.1 NR_002167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017801.mm.1 TC0X00002809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017801.mm.1 NR_002167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017803.mm.1 TC0X00002809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017803.mm.1 NR_002167.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017804.mm.1 TC0X00002809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017804.mm.1 NR_002167.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 640613 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746531 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746531 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0