Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278203_m1
          See other CRXOS GE Assays ›
          Gene Symbol:
          Crxos
          Gene Name:
          cone-rod homeobox, opposite strand
          Gene Aliases:
          AA606869, AY590891, Crxos1, Egam1
          Chromosome Location:
          Chr.7: 15882617 - 15904030 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278203_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crxos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038092.1 3 - 4 245 88
          GenBank mRNA AK149251.1 - 3 - 4 245 88
          AK149256.1 - 3 - 4 245 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB642159.1 - 3 - 4 245 Chr. 7 - 15882617 - 15898722 [+] on Build GRCm38 88
          GenBankmRNA BY742111.1 - 3 - 4 245 Chr. 7 - 15882617 - 15898745 [+] on Build GRCm38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cone-rod homeobox Crx Crx1 -10.6kb Anti-sense Chr. 7 - 15865947 - 15879955 on NCBI Build GRCm38
          Downstream selenoprotein W,
          muscle 1
          Sepw1 selW 19.7kb Anti-sense Chr. 7 - 15917208 - 15922371 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000214.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK149251.1, AK149256.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700001504.mm.1 TC0700000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001504.mm.1 NM_001205274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001504.mm.1 NM_001205274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001504.mm.1 NR_038092.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001504.mm.1 NR_038092.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 TC0700000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 NM_001033638.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 NM_001033638.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 NM_001205274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 NM_001205274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 NR_038092.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001501.mm.1 NR_038092.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 TC0700000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 NM_001033638.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 NM_001033638.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 NM_001205274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 NM_001205274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 NR_038092.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001502.mm.1 NR_038092.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0