Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038348.1 | 2 - 3 | 176 | 75 | ||
GenBank mRNA | DV654203.1 | - | 2 - 3 | 175 | 75 | |
DV662335.1 | - | 2 - 3 | 176 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SOX2 overlapping transcript (non-protein coding) | Sox2ot | B230215L15Rik | -32.2kb | Anti-sense | Chr. 3 - 34560381 - 34677993 on NCBI Build GRCm38 |
Downstream |
predicted gene, 38509 |
Gm38509 | Peril | 58.0kb | Anti-sense | Chr. 3 - 34772075 - 34782346 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() |