Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278271_m1
          See other 9330179D12RIK GE Assays ›
          Gene Symbol:
          9330179D12Rik
          Gene Name:
          RIKEN cDNA 9330179D12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 127149389 - 127212419 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278271_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330179D12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040273.1 2 - 3 405 125
          NR_040274.1 2 - 3 387 125
          GenBank mRNA AK043994.1 - 2 - 3 387 125
          CX238860.1 - 2 - 3 278 125
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB641898.1 - 2 - 3 387 Chr. 6 - 127149389 - 127211781 [+] on Build GRCm38 125

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cyclin D2 Ccnd2 2600016F06Rik,
          AI256817,
          BF642806,
          C86853,
          Vin-1,
          Vin1,
          cD2
          -33.2kb Anti-sense Chr. 6 - 127125162 - 127152188 on NCBI Build GRCm38
          Downstream predicted gene,
          38404
          Gm38404 D130058E03 83.8kb Anti-sense Chr. 6 - 127296186 - 127307967 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001479.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277475 BB749938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277475 AI853374.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277475 AK020388.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688894 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827056 AK029810.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827056 AK129113.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827056 AK147592.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827056 BC041778.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827056 NM_001081057.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87100 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121991 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277475 BB749938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277475 AI853374.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277475 AK020388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328416 AK020388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328416 AK043994.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05168067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05658849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05636634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05401387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05704986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05704988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05018241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05018236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05556478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05789610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05203392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05102652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05721682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24831251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453309_PM_at - -
          Mouse Genome 430 2.0 Array 1453309_at - -
          Mouse Genome 430 2.0 Array 1453309_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 TC0600001479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 NR_040273.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 NR_040273.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 NR_040274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 NR_040274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 NR_040275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012189.mm.1 NR_040275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012183.mm.1 TC0600001479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012183.mm.1 NR_040274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012183.mm.1 NR_040274.1 Refseq
          Mouse Expression Set 430 1453309_at - -
          Mouse Expression Set 430 1453309_PM_at - -
          Murine Genome U74v2 Set 136685_PM_at - -
          Murine Genome U74v2 Set 136685_at - -
          Affymetrix HT Mouse Genome 430B Array 1453309_at - -
          Affymetrix HT Mouse Genome 430B Array 1453309_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 553026 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103140167 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix