Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278288_m1
          See other AI854517 GE Assays ›
          Gene Symbol:
          AI854517
          Gene Name:
          expressed sequence AI854517
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 79500026 - 79534403 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278288_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI854517
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040311.1 4 - 5 527 100
          NR_040312.1 3 - 4 409 100
          NR_040313.1 2 - 3 336 100
          GenBank mRNA BM950091.1 - 4 - 5 433 100
          CX206040.1 - 2 - 3 324 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV458354.1 - 3 - 4 408 Chr. 7 - 79515440 - 79531259 [+] on Build GRCm38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 9-3 Mir9-3 Mirn9-3,
          mir-9-3,
          mmu-mir-9-3
          -22.4kb Sense Chr. 7 - 79505264 - 79505353 on NCBI Build GRCm38
          Downstream Rhesus blood group-associated C glycoprotein Rhcg BB065800 59.0kb Anti-sense Chr. 7 - 79593363 - 79617657 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001124.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AK032343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AK047441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AI853752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AI854517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AK032343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AK047441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AI853752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P301603 AI854517.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06254009 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01557100 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06253952 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06253972 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06253996 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01557084 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01557079 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01557089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06253925 - -
          Whole Mouse Genome G4122A A_51_P301603 AK032343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AK047441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AI853752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AI854517.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AK032343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AK047441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AI853752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301603 AI854517.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125180 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06253952 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01557079 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06253972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06023263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06023271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06312362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06543183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06312357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06023294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06058925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06141356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06103228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06312372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06023282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06116012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06023283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25261628 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1434730_PM_at - -
          Mouse Genome 430 2.0 Array 1434730_at - -
          Mouse Genome 430 2.0 Array 1434730_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101694_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101694_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700009908.mm.1 TC0700001124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009908.mm.1 NR_040312.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009908.mm.1 NR_040312.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 TC0700001124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040311.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040312.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040312.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040313.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040313.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040314.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009919.mm.1 NR_040314.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009907.mm.1 TC0700001124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009907.mm.1 NR_040312.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009907.mm.1 NR_040312.1 Refseq
          Mouse Expression Set 430 1434730_at - -
          Mouse Expression Set 430 1434730_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434730_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434730_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101770093 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0