Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278392_m1
          See other GM20752 GE Assays ›
          Gene Symbol:
          Gm20752
          Gene Name:
          predicted gene, 20752
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 159076364 - 159308080 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278392_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20752
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040750.1 3 - 4 561 79
          GenBank mRNA AK158712.1 - 3 - 4 563 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leucine rich repeat containing 7 Lrrc7 AK122498,
          densin,
          mKIAA1365
          -630.2kb Anti-sense Chr. 3 - 158082888 - 158562221 on NCBI Build GRCm38
          Downstream DEP domain containing 1a Depdc1a 5830484J08Rik,
          Depdc1
          253.1kb Sense Chr. 3 - 159495426 - 159529955 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001586.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548838 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751589 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418999 AK017350.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418999 BC050245.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418999 NM_028973.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138764 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138818 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00804714 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138836 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00804812 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138741 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138813 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138853 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138662 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00804695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138889 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138758 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138778 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138636 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138613 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05138811 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05138636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00804695 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05138802 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05138777 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05138741 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05138836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02826266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02826250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22688687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300012752.mm.1 TC0300001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012752.mm.1 NR_040750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012752.mm.1 NR_040750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012753.mm.1 TC0300001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012753.mm.1 NR_040750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012753.mm.1 NR_040750.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0