Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278610_m1
          See other E330017L17RIK GE Assays ›
          Gene Symbol:
          E330017L17Rik
          Gene Name:
          RIKEN cDNA E330017L17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 129906226 - 129908996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278610_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E330017L17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045190.2 1 - 2 387 75
          GenBank mRNA AK054442.1 - 1 - 2 388 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB666679.1 - 1 - 2 385 Chr. 4 - 129906228 - 129908802 [+] on Build GRCm38 75
          GenBankmRNA BF471383.1 - 1 - 2 390 Chr. 4 - 129906223 - 129908517 [+] on Build GRCm38 75
          GenBankmRNA BB556875.2 - 1 - 2 209 Chr. 4 - 129906404 - 129908997 [+] on Build GRCm38 75
          GenBankmRNA AI596631.1 - 1 - 2 364 Chr. 4 - 129906258 - 129908751 [+] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein tyrosine phosphatase 4a2 Ptp4a2 Prl-2 -56.2kb Sense Chr. 4 - 129820479 - 129850003 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700003M07 gene 1700003M07Rik 51.8kb Sense Chr. 4 - 129960374 - 129965138 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001535.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192633 AK054344.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115682 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192633 AK054344.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04004796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03859284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03706836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03931265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03543981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03893405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03543972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03787147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03706837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03257529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23299074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03257528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444124_PM_a_at - -
          Mouse Genome 430 2.0 Array 1444124_a_at - -
          Mouse Genome 430 2.0 Array 1444124_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR0400011899.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011899.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011904.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011904.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011905.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011905.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011903.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011903.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011897.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011897.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011901.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011901.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011898.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011898.mm.1 NR_045190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011900.mm.1 TC0400001535.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011900.mm.1 NR_045190.2 Refseq
          Mouse Expression Set 430 1444124_PM_a_at - -
          Mouse Expression Set 430 1444124_a_at - -
          Murine Genome U74v2 Set 137376_PM_at - -
          Murine Genome U74v2 Set 137376_at - -
          Affymetrix HT Mouse Genome 430B Array 1444124_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1444124_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 532618 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0