Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278620_m1
          See other AI847159 GE Assays ›
          Gene Symbol:
          AI847159
          Gene Name:
          expressed sequence AI847159
          Gene Aliases:
          OTTMUSG00000015420
          Chromosome Location:
          Chr.2: 129178148 - 129180673 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278620_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI847159
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045264.1 2 - 3 277 64
          GenBank mRNA CK617975.1 - 2 - 3 271 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA EL960245.1 - 2 - 3 276 Chr. 2 - 129178577 - 129180673 [-] on Build GRCm38 64
          GenBankmRNA BI738355.1 - 2 - 3 219 Chr. 2 - 129178205 - 129180615 [-] on Build GRCm38 64
          GenBankmRNA BI738268.1 - 2 - 3 213 Chr. 2 - 129178494 - 129180610 [-] on Build GRCm38 64
          GenBankmRNA CO429158.1 - 2 - 3 234 Chr. 2 - 129178372 - 129180630 [-] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream coiled-coil-helix-coiled-coil-helix domain containing 5 Chchd5 1110027L01Rik,
          AW045710
          -44.0kb Anti-sense Chr. 2 - 129129700 - 129134112 on NCBI Build GRCm38
          Downstream solute carrier family 20,
          member 1
          Slc20a1 AI607883,
          Glvr-1,
          Glvr1
          18.5kb Anti-sense Chr. 2 - 129198773 - 129211615 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004594.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170817 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444095_PM_a_at - -
          Mouse Genome 430 2.0 Array 1444095_a_at - -
          Mouse Genome 430 2.0 Array 1444095_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR0200039355.mm.1 TC0200004594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039355.mm.1 NR_045264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039355.mm.1 NR_045264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039360.mm.1 TC0200004594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039360.mm.1 NR_045264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039360.mm.1 NR_045264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039356.mm.1 TC0200004594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039356.mm.1 NR_045264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039356.mm.1 NR_045264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039358.mm.1 TC0200004594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039358.mm.1 NR_045264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039358.mm.1 NR_045264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039359.mm.1 TC0200004594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039359.mm.1 NR_045264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039359.mm.1 NR_045264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039357.mm.1 TC0200004594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039357.mm.1 NR_045264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039357.mm.1 NR_045264.1 Refseq
          Mouse Expression Set 430 1444095_PM_a_at - -
          Mouse Expression Set 430 1444095_a_at - -
          Murine Genome U74v2 Set 138543_at - -
          Murine Genome U74v2 Set 138543_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444095_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1444095_a_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0