Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278672_m1
          See other 4931403E22RIK GE Assays ›
          Gene Symbol:
          4931403E22Rik
          Gene Name:
          RIKEN cDNA 4931403E22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 26768801 - 26823907 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278672_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4931403E22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045306.1 3 - 4 365 100
          GenBank mRNA AK016426.1 - 3 - 4 366 100
          BY716074.1 - 3 - 4 366 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 20616 Gm20616 -145.4kb Sense Chr. 19 - 26620749 - 26623410 on NCBI Build GRCm38
          Downstream predicted gene 815 Gm815 110.8kb Anti-sense Chr. 19 - 26885925 - 26888647 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001306.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85156 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100683 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03574732 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100683 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09140445 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03574748 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14450072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14183310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14450066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32037382 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900011674.mm.1 TC1900001306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011674.mm.1 NR_045306.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011674.mm.1 NR_045306.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 308766 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104780025 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc