Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278912_m1
          See other 1810020O05RIK GE Assays ›
          Gene Symbol:
          1810020O05Rik
          Gene Name:
          Riken cDNA 1810020O05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 87675593 - 87690847 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278912_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810020O05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045482.1 2 - 3 468 103
          GenBank mRNA AK131825.1 - 2 - 3 469 103
          BY707616.1 - 2 - 3 469 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA E230015B07 gene E230015B07Rik -231.0 Anti-sense Chr. 6 - 87672370 - 87679920 on NCBI Build GRCm38
          Downstream EF hand and coiled-coil domain containing 1 Efcc1 Ccdc48 49.6kb Anti-sense Chr. 6 - 87730820 - 87755912 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002659.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180481 AK131825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180481 AK054052.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178942 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180481 AK131825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180481 AK054052.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05972144 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05972144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05686197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05012980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05057933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05325965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05627163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05841073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05627164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05785312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05707746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05164542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05727334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24617618 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10546051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546050 - -
          Affymetrix Mouse Gene 1.1 ST Array 10546047 - -
          Affymetrix Mouse Clariom S Array PSR0600020681.mm.1 TC0600002659.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020681.mm.1 NR_045482.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020681.mm.1 NR_045482.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020679.mm.1 TC0600002659.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020679.mm.1 NR_045482.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020679.mm.1 NR_045482.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020680.mm.1 TC0600002659.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020680.mm.1 NR_045482.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020680.mm.1 NR_045482.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0