Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335102_m1
          See other 1500004A13RIK GE Assays ›
          Gene Symbol:
          1500004A13Rik
          Gene Name:
          RIKEN cDNA 1500004A13 gene
          Gene Aliases:
          5430404N14Rik, BB132176
          Chromosome Location:
          Chr.3: 88822010 - 88832487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335102_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1500004A13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015498.2 1 - 2 222 81
          GenBank mRNA AK033977.1 - 1 - 2 284 81
          AK036207.1 - 1 - 2 223 81
          AK038592.1 - 1 - 2 223 81
          AK040690.1 - 1 - 2 223 81
          AK078122.1 - 283 81
          AK084105.1 - 1 - 2 223 81
          AK141588.1 - 1 - 2 234 81
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU703566.1 - 1 - 2 206 Chr. 3 - 88824969 - 88832452 [-] on Build GRCm38 81
          GenBankmRNA BB622183.1 - 1 - 2 283 Chr. 3 - 88784424 - 88832547 [-] on Build GRCm38 81
          GenBankmRNA BB652923.1 - 1 - 2 234 Chr. 3 - 88825006 - 88832498 [-] on Build GRCm38 81
          GenBankmRNA BF471983.1 - 1 - 2 112 Chr. 3 - 88821475 - 88832377 [-] on Build GRCm38 81
          GenBankmRNA BB632433.1 - 1 - 2 223 Chr. 3 - 88825024 - 88832487 [-] on Build GRCm38 81
          GenBankmRNA BB636604.1 - 1 - 2 223 Chr. 3 - 88825129 - 88832487 [-] on Build GRCm38 81
          GenBankmRNA BB628999.1 - 1 - 2 223 Chr. 3 - 88825003 - 88832487 [-] on Build GRCm38 81
          GenBankmRNA BB657396.1 - 1 - 2 223 Chr. 3 - 88824991 - 88832487 [-] on Build GRCm38 81
          GenBankmRNA CX240898.1 - 1 - 2 264 Chr. 3 - 88824914 - 88832529 [-] on Build GRCm38 81
          GenBankmRNA BY726972.1 - 1 - 2 283 Chr. 3 - 88825084 - 88832547 [-] on Build GRCm38 81
          GenBankmRNA W08215.1 - 1 - 2 196 Chr. 3 - 88825244 - 88832452 [-] on Build GRCm38 81
          GenBankmRNA CB848283.1 - 1 - 2 145 Chr. 3 - 88811223 - 88832410 [-] on Build GRCm38 81
          GenBankmRNA CA321648.1 - 1 - 2 195 Chr. 3 - 88824482 - 88832458 [-] on Build GRCm38 81
          GenBankmRNA BY065836.1 - 1 - 2 223 Chr. 3 - 88825272 - 88832487 [-] on Build GRCm38 81
          GenBankmRNA CF724755.1 - 1 - 2 198 Chr. 3 - 88824886 - 88832463 [-] on Build GRCm38 81
          GenBankmRNA BB625839.1 - 1 - 2 283 Chr. 3 - 88825045 - 88832547 [-] on Build GRCm38 81

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream gon-4-like pseudogene 5830417I10Rik 3632445O20Rik,
          AA387005
          Anti-sense Chr. 3 - 88777057 - 88829371 on NCBI Build GRCm38
          Downstream misato homolog 1 (Drosophila) Msto1 BC008103,
          mst
          77.1kb Sense Chr. 3 - 88909616 - 88913950 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0