Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335133_m1
          See other 4933439C10RIK GE Assays ›
          Gene Symbol:
          4933439C10Rik
          Gene Name:
          RIKEN cDNA 4933439C10 gene
          Gene Aliases:
          2610028M21Rik
          Chromosome Location:
          Chr.11: 59505685 - 59511067 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335133_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933439C10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015585.2 4 - 5 1527 187
          GenBank mRNA AK017115.1 - 4 - 5 1528 187
          AK163429.1 - 4 - 5 1523 187
          AK169756.1 - 4 - 5 1557 187
          BC065402.1 - 3 - 4 233 187
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY762673.1 - 3 - 4 342 Chr. 11 - 59507190 - 59511067 [+] on Build GRCm38 187
          GenBankmRNA AI036408.1 - 3 - 4 219 Chr. 11 - 59507641 - 59511012 [+] on Build GRCm38 187
          GenBankmRNA CJ317149.1 - 1 - 2 95 Chr. 11 - 59509654 - 59511070 [+] on Build GRCm38 187
          GenBankmRNA BP760795.1 - 1 - 2 Chr. 11 - 59507840 - 59511073 [-] on Build GRCm38 187
          GenBankmRNA EH098200.1 - 2 - 3 201 Chr. 11 - 59507842 - 59511069 [+] on Build GRCm38 187
          GenBankmRNA AI429744.1 - 1 - 2 Chr. 11 - 59507887 - 59511073 [-] on Build GRCm38 187
          GenBankmRNA EH099787.1 - 3 - 4 342 Chr. 11 - 59507189 - 59510930 [+] on Build GRCm38 187
          GenBankmRNA BE947806.1 - 1 - 2 Chr. 11 - 59509708 - 59511073 [-] on Build GRCm38 187
          GenBankmRNA BE136486.1 - 1 - 2 Chr. 11 - 59509649 - 59511073 [-] on Build GRCm38 187
          GenBankmRNA BB802259.1 - 1 - 2 60 Chr. 11 - 59509689 - 59511067 [+] on Build GRCm38 187
          GenBankmRNA BE133382.1 - 3 - 4 233 Chr. 11 - 59507283 - 59510940 [+] on Build GRCm38 187
          GenBankmRNA EH102917.1 - 3 - 4 342 Chr. 11 - 59507189 - 59511035 [+] on Build GRCm38 187

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger with KRAB and SCAN domains 17 Zkscan17 Nizp1,
          Zfp496,
          Znf496
          -3.0kb Anti-sense Chr. 11 - 59485520 - 59506640 on NCBI Build GRCm38
          Downstream NLR family,
          pyrin domain containing 3
          Nlrp3 AGTAVPRL,
          AII/AVP,
          Cias1,
          FCAS,
          FCU,
          MWS,
          Mmig1,
          NALP3,
          Pypaf1
          30.5kb Sense Chr. 11 - 59541569 - 59566956 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000725.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2408651 AK051100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 AK169756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 BC065402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 AK163429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 AK017115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 NR_015585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 AK169756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 BC065402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 AK163429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 AK017115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408749 NR_015585.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413818 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137920 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2408651 AK051100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 AK169756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 BC065402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 AK163429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 AK017115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 NR_015585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 AK169756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 BC065402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 AK163429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 AK017115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408749 NR_015585.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137920 - -
          Whole Mouse Genome G4122A A_51_P408749 AK169756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 BC065402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 AK163429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 AK017115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 NR_015585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 AK169756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 BC065402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 AK163429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 AK017115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408749 NR_015585.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137920 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376509 - -
          Affymetrix HT MG-430 PM Array Plate 1438437_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1456408_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1452696_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376507 - -
          Mouse Genome 430 2.0 Array 1452696_a_at - -
          Mouse Genome 430 2.0 Array 1438437_a_at - -
          Mouse Genome 430 2.0 Array 1456408_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456408_x_at - -
          Mouse Genome 430 2.0 Array 1452696_PM_a_at - -
          Mouse Genome 430 2.0 Array 1438437_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR1100005783.mm.1 TC1100000725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005783.mm.1 NR_015585.3 Refseq
          Mouse Expression Set 430 1438437_a_at - -
          Mouse Expression Set 430 1456408_PM_x_at - -
          Mouse Expression Set 430 1452696_a_at - -
          Mouse Expression Set 430 1452696_PM_a_at - -
          Mouse Expression Set 430 1438437_PM_a_at - -
          Mouse Expression Set 430 1456408_x_at - -
          Murine Genome U74v2 Set 138081_at - -
          Murine Genome U74v2 Set 138081_PM_at - -
          Murine Genome U74v2 Set 171251_PM_at - -
          Murine Genome U74v2 Set 171251_at - -
          Affymetrix HT Mouse Genome 430B Array 1438437_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1456408_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1438437_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1452696_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1452696_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1456408_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106450195 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix