Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335844_m1
          See other F420014N23RIK GE Assays ›
          Gene Symbol:
          F420014N23Rik
          Gene Name:
          RIKEN cDNA F420014N23 gene
          Gene Aliases:
          ENSMUSG00000074658
          Chromosome Location:
          Chr.10: 127195249 - 127202643 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335844_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F420014N23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045715.1 3 - 4 433 70
          NR_045716.1 2 - 3 277 70
          GenBank mRNA AK077682.1 - 3 - 4 434 70
          AK165234.1 - 2 - 3 278 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV304361.2 - 3 - 4 427 Chr. 10 - 127195256 - 127200750 [+] on Build GRCm38 70
          GenBankmRNA BY732386.1 - 2 - 3 278 Chr. 10 - 127195249 - 127200925 [+] on Build GRCm38 70
          GenBankmRNA BB619271.1 - 3 - 4 434 Chr. 10 - 127195249 - 127200728 [+] on Build GRCm38 70
          GenBankmRNA AI466698.1 - 2 - 3 317 Chr. 10 - 127195266 - 127200701 [+] on Build GRCm38 70
          GenBankmRNA AI465162.1 - 1 - 2 Chr. 10 - 127195448 - 127200752 [-] on Build GRCm38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphatidylinositol-5-phosphate 4-kinase,
          type II,
          gamma
          Pip4k2c Pip5k2c Anti-sense Chr. 10 - 127197067 - 127211644 on NCBI Build GRCm38
          Downstream kinesin family member 5A Kif5a D10Bwg0738e,
          Khc,
          Kif5,
          Kns,
          mKIAA4086
          23.1kb Anti-sense Chr. 10 - 127225695 - 127263363 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001553.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK165234.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2221484 AK165234.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2221484 AK165234.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P811856 AK165234.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10366865 - -
          Affymetrix Mouse Gene 1.1 ST Array 10366864 - -
          Affymetrix Mouse Clariom S Array PSR1000011027.mm.1 TC1000001553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011027.mm.1 NR_045715.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011027.mm.1 NR_045715.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011027.mm.1 NR_045716.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011027.mm.1 NR_045716.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0