Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335873_g1
          See other 1810034E14RIK GE Assays ›
          Gene Symbol:
          1810034E14Rik
          Gene Name:
          RIKEN cDNA 1810034E14 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 64248700 - 64268145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335873_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810034E14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045798.1 2 - 3 680 96
          GenBank mRNA AK002708.1 - 2 - 3 328 96
          AK143964.1 - 2 - 3 635 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY703213.1 - 2 - 3 328 Chr. 13 - 64249052 - 64267496 [+] on Build GRCm38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream hyaluronic acid binding protein 4 Habp4 4933413D03Rik,
          4933428J01Rik
          -79.8kb Sense Chr. 13 - 64161866 - 64186537 on NCBI Build GRCm38
          Downstream AhpC/TSA antioxidant enzyme domain containing 1 Aaed1 1110018J18Rik,
          AI849003
          23.7kb Anti-sense Chr. 13 - 64291836 - 64312710 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000779.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1700000027.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the AK143964.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660047 AK002708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660047 AK007690.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660047 AK143964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660047 AK002708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660047 AK007690.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P660047 AK143964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660047 AK002708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660047 AK007690.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660047 AK143964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660047 AK002708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660047 AK007690.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P660047 AK143964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660047 AK002708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660047 AK007690.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660047 AK143964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660047 AK002708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660047 AK007690.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660047 AK143964.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140777 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430657_PM_at - -
          Mouse Genome 430 2.0 Array 1430657_at - -
          Mouse Genome 430 2.0 Array 1430657_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300005163.mm.1 TC1300000779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005163.mm.1 NR_045798.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005163.mm.1 NR_045798.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005164.mm.1 TC1300000779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005164.mm.1 NR_045798.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005164.mm.1 NR_045798.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005165.mm.1 TC1300000779.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300005165.mm.1 NR_045798.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005165.mm.1 NR_045798.1 Refseq
          Mouse Expression Set 430 1430657_PM_at - -
          Mouse Expression Set 430 1430657_at - -
          Affymetrix HT Mouse Genome 430B Array 1430657_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430657_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-746d97c75b-2cnrv:80/100.66.135.11:80.
          git-commit: a7e96c5202fecf029dfaed736aa540acb41e6c9b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/PDP_Cmrc_Card_Notification_Changes