Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335885_m1
          See other 5430440P10RIK GE Assays ›
          Gene Symbol:
          5430440P10Rik
          Gene Name:
          RIKEN cDNA 5430440P10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 105427559 - 105437716 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335885_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430440P10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045859.1 1 - 2 120 176

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Nedd4 family interacting protein 2 Ndfip2 0710001O20Rik,
          2810436B12Rik,
          9130207N19Rik,
          N4wbp5a,
          mKIAA1165
          -118.3kb Sense Chr. 14 - 105258673 - 105309298 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930449E01 gene 4930449E01Rik 65.3kb Sense Chr. 14 - 105498788 - 105505629 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001202.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23270 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160952 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160952 - -
          Whole Mouse Genome G4122A A_51_P509418 AK017418.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109894 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11761918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29877010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11275149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29877003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11651807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11761920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29877001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11761914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11761923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11804546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29877004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11761900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29877012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11761916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29876999 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432388_PM_at - -
          Mouse Genome 430 2.0 Array 1432388_at - -
          Mouse Genome 430 2.0 Array 1432388_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400009446.mm.1 TC1400001202.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009446.mm.1 NR_045859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009446.mm.1 NR_045859.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009444.mm.1 TC1400001202.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009444.mm.1 NR_045859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009444.mm.1 NR_045859.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009448.mm.1 TC1400001202.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009448.mm.1 NR_045859.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009448.mm.1 NR_045859.1 Refseq
          Mouse Expression Set 430 1432388_PM_at - -
          Mouse Expression Set 430 1432388_at - -
          Affymetrix HT Mouse Genome 430B Array 1432388_at - -
          Affymetrix HT Mouse Genome 430B Array 1432388_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 721257 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104760364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0