Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335950_m1
          See other GM10445 GE Assays ›
          Gene Symbol:
          Gm10445
          Gene Name:
          predicted gene 10445
          Gene Aliases:
          ENSMUSG00000072956
          Chromosome Location:
          Chr.6: 84412942 - 84423838 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335950_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10445
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046063.1 2 - 3 355 80
          GenBank mRNA AK144571.1 - 2 - 3 356 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY733904.1 - 2 - 3 353 Chr. 6 - 84413031 - 84423836 [-] on Build GRCm38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dysferlin Dysf 2310004N10Rik,
          AI604795,
          D6Pas3
          -209.3kb Anti-sense Chr. 6 - 84008316 - 84211060 on NCBI Build GRCm38
          Downstream cytochrome P450,
          family 26,
          subfamily b,
          polypeptide 1
          Cyp26b1 CP26,
          P450RAI-2
          149.8kb Sense Chr. 6 - 84571414 - 84593908 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002590.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140712 AK144571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140712 AK144571.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01372413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05446027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05051412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05688803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05256373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05494475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05428434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05169654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05091617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05256382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05744857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05147648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05745533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05304171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05051406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05446022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05253887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05446025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05169655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05745539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05185736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05759775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05446018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05147619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05033801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05497084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05446029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05253884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05253885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05304177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05185737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05051414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05253886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05147626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05582277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05744856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05147620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05446019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05135556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05239854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05304175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05304174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05632176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05494477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05135554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05660078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24599313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600020131.mm.1 TC0600002590.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020131.mm.1 NR_046063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020131.mm.1 NR_046063.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020130.mm.1 TC0600002590.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020130.mm.1 NR_046063.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020130.mm.1 NR_046063.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0