Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04410280_m1
          See other TYSND1 GE Assays ›
          Gene Symbol:
          Tysnd1
          Gene Name:
          trypsin domain containing 1
          Gene Aliases:
          1300019N10Rik
          Chromosome Location:
          Chr.10: 61695497 - 61702773 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04410280_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tysnd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073587.1 1 - 2 44 65
          GenBank mRNA BE847683.1 - 1 - 2 44 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SAR1 gene homolog A (S. cerevisiae) Sar1a 1600019H17Rik,
          Sara,
          Sara1
          -2.3kb Sense Chr. 10 - 61680304 - 61693297 on NCBI Build GRCm38
          Downstream apoptosis-inducing factor,
          mitochondrion-associated 2
          Aifm2 5430437E11Rik,
          Amid,
          D730001I10Rik,
          PRG3
          12.5kb Sense Chr. 10 - 61715263 - 61739260 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000573.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein processing
          regulation of fatty acid beta-oxidation
          protein homooligomerization
          protease binding
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          identical protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 TC1000000573.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 NM_001272090.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 NM_001272091.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 NM_001272092.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 NM_027912.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 NM_027912.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003883.mm.1 NR_073587.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 TC1000000573.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 NM_001272090.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 NM_001272091.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 NM_001272092.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 NM_027912.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 NM_027912.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003888.mm.1 NR_073587.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 TC1000000573.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 NM_001272090.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 NM_001272091.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 NM_001272092.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 NM_027912.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 NM_027912.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003889.mm.1 NR_073587.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 820860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0