Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5830444B04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102283.1 | 1 - 2 | 67 | 68 | ||
NR_102284.1 | 1 - 2 | 67 | 68 | |||
NR_102287.1 | 1 - 2 | 67 | 68 | |||
GenBank mRNA | AK167606.1 | - | 64 | 68 | ||
BY737327.1 | - | 1 - 2 | 64 | 68 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cilia and flagella associated protein 74 | Cfap74 |
2010015L04Rik, A530045M11 |
Anti-sense | Chr. 4 - 155398208 - 155466823 on NCBI Build GRCm38 | |
Downstream | transmembrane protein 52 | Tmem52 | 2310043I08Rik | 47.4kb | Anti-sense | Chr. 4 - 155469114 - 155470858 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() |