Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04411092_g1
          See other HNRNPA2B1 GE Assays ›
          Gene Symbol:
          Hnrnpa2b1
          Gene Name:
          heterogeneous nuclear ribonucleoprotein A2/B1
          Gene Aliases:
          9130414A06Rik, Hnrpa2, Hnrpa2b1, hnrnp-A
          Chromosome Location:
          Chr.6: 51460434 - 51470492 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04411092_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnpa2b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104468.1 10 - 11 1282 79
          GenBank mRNA AK049735.1 - 1 - 2 1267 79
          AK089012.1 - 10 - 11 1233 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA185991.1 - 1 - 2 64 Chr. 6 - 51461121 - 51461982 [-] on Build GRCm38 79
          GenBankmRNA AA286353.1 - 1 - 2 124 Chr. 6 - 51461123 - 51462042 [-] on Build GRCm38 79
          GenBankmRNA CB523961.1 - 1 - 2 148 Chr. 6 - 51461121 - 51462058 [-] on Build GRCm38 79
          GenBankmRNA BE849229.1 - 1 - 2 43 Chr. 6 - 51461125 - 51461961 [-] on Build GRCm38 79
          GenBankmRNA BF661658.1 - 1 - 2 Chr. 6 - 51461121 - 51462017 [+] on Build GRCm38 79
          GenBankmRNA AV570772.1 - 3 - 4 349 Chr. 6 - 51461260 - 51464209 [-] on Build GRCm38 79
          GenBankmRNA BM229222.1 - 1 - 2 Chr. 6 - 51461124 - 51462112 [+] on Build GRCm38 79
          GenBankmRNA BE332292.1 - 1 - 2 414 Chr. 6 - 51461380 - 51462332 [-] on Build GRCm38 79
          GenBankmRNA C76903.1 - Chr. - - [-] on Build 79
          GenBankmRNA DT911660.1 - 1 - 2 Chr. 6 - 51461361 - 51462409 [+] on Build GRCm38 79
          GenBankmRNA AA174432.1 - 1 - 2 313 Chr. 6 - 51461250 - 51462231 [-] on Build GRCm38 79
          GenBankmRNA AW554484.1 - 1 - 2 Chr. 6 - 51461124 - 51462085 [+] on Build GRCm38 79
          GenBankmRNA AW907586.1 - 1 - 2 Chr. 6 - 51461294 - 51461979 [+] on Build GRCm38 79
          GenBankmRNA BQ177318.1 - 1 - 2 476 Chr. 6 - 51461240 - 51462394 [-] on Build GRCm38 79
          GenBankmRNA BQ102179.1 - 1 - 2 165 Chr. 6 - 51461121 - 51462083 [-] on Build GRCm38 79
          GenBankmRNA CR758644.1 - 1 - 2 Chr. 6 - 51461360 - 51462427 [+] on Build GRCm38 79
          GenBankmRNA EH100464.1 - 148 Chr. - - [-] on Build 79
          GenBankmRNA CB842739.2 - 61 Chr. - - [-] on Build 79
          GenBankmRNA AW907986.1 - 1 - 2 305 Chr. 6 - 51461310 - 51462191 [-] on Build GRCm38 79
          GenBankmRNA BB425228.2 - 1 - 2 304 Chr. 6 - 51461147 - 51462222 [-] on Build GRCm38 79
          GenBankmRNA BF453009.1 - 1 - 2 325 Chr. 6 - 51461326 - 51462243 [-] on Build GRCm38 79
          GenBankmRNA AA122806.1 - 2 - 3 169 Chr. 6 - 51461130 - 51463442 [-] on Build GRCm38 79
          GenBankmRNA BY763450.1 - 3 - 4 367 Chr. 6 - 51461124 - 51464213 [-] on Build GRCm38 79
          GenBankmRNA BE311340.1 - 1 - 2 Chr. 6 - 51461175 - 51462191 [+] on Build GRCm38 79
          GenBankmRNA BM114941.1 - 1 - 2 Chr. 6 - 51461124 - 51462128 [+] on Build GRCm38 79
          GenBankmRNA BI790966.1 - 1 - 2 Chr. 6 - 51461122 - 51462147 [+] on Build GRCm38 79
          GenBankmRNA AA107649.1 - 2 - 3 151 Chr. 6 - 51461157 - 51462104 [-] on Build GRCm38 79
          GenBankmRNA BF453050.1 - 1 - 2 280 Chr. 6 - 51461237 - 51462198 [-] on Build GRCm38 79
          GenBankmRNA BM201112.1 - 1 - 2 Chr. 6 - 51461140 - 51463498 [+] on Build GRCm38 79
          GenBankmRNA CK380653.1 - 4 - 5 402 Chr. 6 - 51461224 - 51464469 [-] on Build GRCm38 79
          GenBankmRNA AV583518.1 - 1 - 2 111 Chr. 6 - 51461121 - 51462029 [-] on Build GRCm38 79
          GenBankmRNA AI449004.1 - 1 - 2 205 Chr. 6 - 51461237 - 51462123 [-] on Build GRCm38 79
          GenBankmRNA AU257266.1 - 1 - 2 53 Chr. 6 - 51461166 - 51461971 [-] on Build GRCm38 79
          GenBankmRNA C88150.1 - Chr. - - [-] on Build 79
          GenBankmRNA DV046474.1 - 8 - 9 786 Chr. 6 - 51461303 - 51466545 [-] on Build GRCm38 79
          GenBankmRNA CK031504.1 - 4 - 5 421 Chr. 6 - 51461121 - 51464505 [-] on Build GRCm38 79
          GenBankmRNA BQ101905.1 - 1 - 2 Chr. 6 - 51461294 - 51462114 [+] on Build GRCm38 79
          GenBankmRNA BE292533.1 - 1 - 2 287 Chr. 6 - 51461194 - 51462191 [-] on Build GRCm38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nuclear factor,
          erythroid derived 2,
          like 3
          Nfe2l3 Nrf3 -1.7kb Anti-sense Chr. 6 - 51432670 - 51458768 on NCBI Build GRCm38
          Downstream chromobox 3 Cbx3 HP1g,
          M32
          8.6kb Anti-sense Chr. 6 - 51470616 - 51483704 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002265.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          mRNA processing
          RNA splicing
          negative regulation of mRNA splicing, via spliceosome
          RNA transport
          nucleotide binding
          regulatory region RNA binding
          nucleic acid binding
          RNA binding
          single-stranded telomeric DNA binding
          pre-mRNA intronic binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600017751.mm.1 TC0600002265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017751.mm.1 NM_016806.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017751.mm.1 NM_182650.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017751.mm.1 NR_104468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017754.mm.1 TC0600002265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017754.mm.1 NM_016806.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017754.mm.1 NM_182650.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017754.mm.1 NR_104468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017752.mm.1 TC0600002265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017752.mm.1 NM_016806.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017752.mm.1 NM_182650.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017752.mm.1 NR_104468.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 391014 - -
          ABI Mouse Genome Survey Microarray 704801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0