Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04412128_m1
          See other TMEM161A GE Assays ›
          Gene Symbol:
          Tmem161a
          Gene Name:
          transmembrane protein 161A
          Gene Aliases:
          AI428876, BB161850, BC021367
          Chromosome Location:
          Chr.8: 70172329 - 70183681 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04412128_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem161a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293797.1 NP_001280726.1 4 - 5 372 79
          NM_145597.4 NP_663572.1 5 - 6 476 79
          NR_122028.1 5 - 6 476 79
          GenBank mRNA AK049979.1 - 5 - 6 475 79
          AK082758.1 - 5 - 6 472 79
          AK082916.1 - 4 - 5 373 79
          AK088569.1 - 4 - 5 362 79
          AK147962.1 - 5 - 6 469 79
          BC021367.1 - 5 - 6 476 5030434 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY746737.1 - 4 - 5 362 Chr. 8 - 70172419 - 70180785 [+] on Build GRCm38 79
          GenBankmRNA CF163227.1 - 1 - 2 122 Chr. 8 - 70177490 - 70180995 [+] on Build GRCm38 79
          GenBankmRNA BF237404.1 - 3 - 4 305 Chr. 8 - 70176879 - 70180831 [+] on Build GRCm38 79
          GenBankmRNA BI695929.1 - 4 - 5 459 Chr. 8 - 70174302 - 70180808 [+] on Build GRCm38 79
          GenBankmRNA BB867795.1 - 4 - 5 404 Chr. 8 - 70172419 - 70178975 [+] on Build GRCm38 79
          GenBankmRNA BF136392.1 - 4 - 5 444 Chr. 8 - 70174303 - 70178982 [+] on Build GRCm38 79
          GenBankmRNA BG288530.1 - 416 Chr. - - [-] on Build 79
          GenBankmRNA BQ554153.1 - 1 - 2 122 Chr. 8 - 70177490 - 70181049 [+] on Build GRCm38 79
          GenBankmRNA CA752064.1 - 4 - 5 452 Chr. 8 - 70174302 - 70180779 [+] on Build GRCm38 79
          GenBankmRNA BG972253.1 - 444 Chr. - - [-] on Build 79
          GenBankmRNA BY741187.1 - 5 - 6 466 Chr. 8 - 70172418 - 70180726 [+] on Build GRCm38 79
          GenBankmRNA BB864447.1 - 2 - 3 212 Chr. 8 - 70177289 - 70180723 [+] on Build GRCm38 78
          GenBankmRNA BY224742.1 - 4 - 5 364 Chr. 8 - 70172416 - 70178935 [+] on Build GRCm38 79
          GenBankmRNA CA564277.1 - 1 - 2 122 Chr. 8 - 70177490 - 70180843 [+] on Build GRCm38 79
          GenBankmRNA CK793590.1 - 4 - 5 462 Chr. 8 - 70174302 - 70180787 [+] on Build GRCm38 79
          GenBankmRNA BE288806.1 - 4 - 5 426 Chr. 8 - 70174322 - 70178965 [+] on Build GRCm38 79
          GenBankmRNA BI112590.1 - 5 - 6 465 Chr. 8 - 70172419 - 70180758 [+] on Build GRCm38 79
          GenBankmRNA CA543170.1 - 1 - 2 122 Chr. 8 - 70177490 - 70180795 [+] on Build GRCm38 79
          GenBankmRNA CD560194.1 - 1 - 2 122 Chr. 8 - 70177490 - 70180729 [+] on Build GRCm38 79
          GenBankmRNA CF167225.1 - 1 - 2 122 Chr. 8 - 70177490 - 70180800 [+] on Build GRCm38 79
          GenBankmRNA BQ946775.1 - 4 - 5 357 Chr. 8 - 70172423 - 70181171 [+] on Build GRCm38 79
          GenBankmRNA BG061485.2 - 1 - 2 122 Chr. 8 - 70177490 - 70180790 [+] on Build GRCm38 79
          GenBankmRNA BF579666.1 - 192 Chr. - - [-] on Build 79
          GenBankmRNA BI655196.1 - 3 - 4 400 Chr. 8 - 70176844 - 70181021 [+] on Build GRCm38 79
          GenBankmRNA CK617649.1 - 4 - 5 455 Chr. 8 - 70174302 - 70179019 [+] on Build GRCm38 79
          GenBankmRNA BE304005.1 - 4 - 5 441 Chr. 8 - 70174305 - 70179018 [+] on Build GRCm38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream myocyte enhancer factor 2B Mef2b AI451606 -10.0kb Sense Chr. 8 - 70152778 - 70167488 on NCBI Build GRCm38
          Downstream solute carrier family 25,
          member 42
          Slc25a42 2900084M01Rik 5.3kb Anti-sense Chr. 8 - 70184340 - 70212281 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK082916.1, NM_001293797.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem161a

          Gene Ontology Categories:

          Function(s) Process(es)

          response to retinoic acid
          cellular response to oxidative stress
          cellular response to UV
          positive regulation of DNA repair
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          molecular_function

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
          git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0