Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101109335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965171.1 | XP_027820972.1 | 4 - 5 | 452 | 82 |
Set Membership: |
![]() ![]() ![]() |