Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Oa04840480_m1
See other LOC101104231 GE Assays ›
Gene Symbol:
LOC101104231
Gene Name:
calcium-dependent secretion activator 2
Gene Aliases:
CADPS2
Chromosome Location:
Chr.4: 94821737 - 95396545 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID
Oa04840480_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: LOC101104231
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004008016.4
XP_004008065.1
7 - 8
1605
69
XM_004008017.4
XP_004008066.1
7 - 8
1606
69
XM_004008018.4
XP_004008067.1
7 - 8
1605
69
XM_004008019.4
XP_004008068.1
7 - 8
1606
69
XM_012177037.3
XP_012032427.1
7 - 8
1606
69
XM_012177040.3
XP_012032430.1
7 - 8
1606
69
XM_012177041.3
XP_012032431.1
7 - 8
1606
69
XM_012177042.3
XP_012032432.1
7 - 8
1607
69
XM_012177043.3
XP_012032433.1
7 - 8
1606
69
XM_012177044.3
XP_012032434.1
7 - 8
1606
69
XM_012177046.3
XP_012032436.1
7 - 8
1608
69
XM_012177047.3
XP_012032437.1
7 - 8
1606
69
XM_012177049.3
XP_012032439.1
7 - 8
1605
69
XM_012177051.3
XP_012032441.1
7 - 8
1610
69
XM_012177052.3
XP_012032442.1
7 - 8
1605
69
XM_012177053.3
XP_012032443.1
7 - 8
1605
69
XM_012177054.3
XP_012032444.1
7 - 8
1605
69
XM_027968766.1
XP_027824567.1
7 - 8
1605
69
XM_027968767.1
XP_027824568.1
7 - 8
1605
69
XM_027968768.1
XP_027824569.1
7 - 8
1605
69
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Ovis aries
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0