Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012176900.3 | XP_012032290.3 | 1 - 2 | 889 | 104 | |
XM_027968662.1 | XP_027824463.1 | 2 - 3 | 685 | 104 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |