Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101121595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968820.1 | XP_027824621.1 | 4 - 5 | 1533 | 106 | |
XM_027968822.1 | XP_027824623.1 | 4 - 5 | 1535 | 106 | ||
XM_027968823.1 | XP_027824624.1 | 4 - 5 | 606 | 106 | ||
XM_027968825.1 | XP_027824626.1 | 4 - 5 | 647 | 106 | ||
XM_027968826.1 | XP_027824627.1 | 4 - 5 | 1530 | 106 | ||
XM_027968827.1 | XP_027824628.1 | 4 - 5 | 634 | 106 | ||
XM_027968828.1 | XP_027824629.1 | 4 - 5 | 626 | 106 |
Set Membership: |
![]() ![]() ![]() |