Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965426.1 | XP_027821227.1 | 2 - 3 | 556 | 54 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |