Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082175.2 | NP_001075644.1 | 2 | 546 | 103 | |
GenBank mRNA | AY630339.1 | - | 2 | 548 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |