Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017345345.1 | XP_017200834.1 | 1 - 2 | 397 | 85 | |
XM_017345346.1 | XP_017200835.1 | 1 - 2 | 397 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() |