Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100340255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017347053.1 | XP_017202542.1 | 7 - 8 | 3951 | 91 | |
XM_017347054.1 | XP_017202543.1 | 6 - 7 | 3058 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() |